missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleic hydrazide, 97%

Catalog No. p-7025484
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2097506 5 g
AAB2097514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2097506 Supplier Thermo Scientific Chemicals Supplier No. B2097506
Only null left

CAS: 5436-01-1 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5436-01-1
MDL Number MFCD00234108

Specifications

Melting Point 216°C to 219°C
Molecular Formula C10H13NO2
Quantity 5 g
Synonym 2-amino-2-phenylbutyric acid, 2-ammonio-2-phenylbutanoate, benzeneacetic acid, a-amino-a-ethyl, 1-2-amino-2-phenylbutyric acid, aminophenylbutyric acid, acmc-20ao5m, 4-sulfobenzoic anhydride, 2-ethyl-2-phenylglycine, 2-amino-2-phenulbutyric acid, 2-amino-2-phenyl-butyric acid
InChI Key UBXUDSPYIGPGGP-UHFFFAOYNA-N
SMILES CCC(C1=CC=CC=C1)(C(=O)O)N
IUPAC Name 2-amino-2-phenylbutanoic acid
Molecular Weight (g/mol) 179.22
PubChem CID 89317
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material N-Methylmaleic hydrazide
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only