missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleic Acid Monoamide 98.0+%, TCI America™

Catalog No. M100025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M100025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M100025G Supplier TCI America Supplier No. M100025G
Only null left

Chemical Identifiers

CAS 6936-48-7
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00014012
InChI Key DFQUBYCHLQAFOW-IHWYPQMZSA-N
Synonym n-methylmaleamic acid, z-4-methylamino-4-oxobut-2-enoic acid, n-methylmaleic acid monoamide, 2z-3-methylcarbamoyl prop-2-enoic acid, maleamic acid, n-methyl, 2z-4-methylamino-4-oxo-2-butenoic acid, 3-methylaminocarbonyl acrylic acid, n-methylmaleamic, hifl@nafr vvfzafh, n-methyl maleamic acid
PubChem CID 5354909
IUPAC Name (Z)-4-(methylamino)-4-oxobut-2-enoic acid
SMILES CNC(=O)C=CC(=O)O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 129.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylmaleic Acid Monoamide