missing translation for 'onlineSavingsMsg'
Learn More

Anhydride N-méthylisatoïque 98,0+%, TCI America™

Numéro de catalogue. M074325G
Modifier l'affichage
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M074325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M074325G Fournisseur TCI America Code fournisseur M074325G

Identifiants chimiques

CAS 10328-92-4
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00006815
InChI Key KJMRWDHBVCNLTQ-UHFFFAOYSA-N
Synonym n-methylisatoic anhydride, 1-methyl-1h-benzo d 1,3 oxazine-2,4-dione, n-methyl ia, 1-methyl-2h-3,1-benzoxazine-2,4 1h-dione, 2h-3,1-benzoxazine-2,4 1h-dione, 1-methyl, n-methyl-n-carboxyanthranilic anhydride, methyl-n-carboxyanthranilic anhydride, 1-methylbenzo d 1,3-oxazine-2,4-dione, n-methyl isotoic anhydride, 1-methyl-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione
PubChem CID 25160
ChEBI CHEBI:59054
IUPAC Name 1-méthyl-3,1-benzoxazine-2,4-dione
SMILES CN1C2=CC=CC=C2C(=O)OC1=O

Spécifications

Melting Point 179°C
Color Blanc-jaune
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material N-Methylisatoic Anhydride
RTECSNumber DM3140000
TSCA Yes