missing translation for 'onlineSavingsMsg'
Learn More

N-Methylhydroxylamine hydrochloride, 98%

Catalog No. AAL0220206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0220206 5 g
AAL0220214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0220206 Supplier Thermo Scientific Chemicals Supplier No. L0220206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4229-44-1 | CH6ClNO | 83.52 g/mol

N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and air.

Chemical Identifiers

CAS 4229-44-1
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012597
InChI Key RGZRSLKIOCHTSI-UHFFFAOYSA-N
Synonym n-methylhydroxylamine hydrochloride, methylhydroxylammonium chloride, methylhydroxylamine hydrochloride, n-methylhydroxylamine hcl, unii-mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride, n-methylhydroxylammonium chloride, mgw04sj1lz, methanamine, n-hydroxy-, hydrochloride 1:1, menhoh.hcl
PubChem CID 77906
IUPAC Name N-methylhydroxylamine;hydrochloride
SMILES [H+].[Cl-].CNO

Specifications

Melting Point 80°C to 86°C
Quantity 5 g
Beilstein 3541409
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 83.52
Percent Purity 98%
Chemical Name or Material N-Methylhydroxylamine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-181-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only