missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine 97.0+%, TCI America™

Catalog No. M05185ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05185ML 5 mL
M051825ML 25 mL
M0518100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M05185ML Supplier TCI America Supplier No. M05185ML

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name (2-azaniumylethyl)(methyl)azanium
SMILES C[NH2+]CC[NH3+]

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 mL
UN Number 2734
Formula Weight 74.13
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N-Methylethylenediamine
RTECSNumber KV5250500
TSCA Yes