missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine, 95%

Catalog No. AAA1216522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1216522 100 g
AAA1216506 5 g
AAA1216514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1216522 Supplier Thermo Scientific Chemicals Supplier No. A1216522

CAS: 109-81-9 | C3H12N2 | 76.14 g/mol

N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name N'-methylethane-1,2-diamine
SMILES C[NH2+]CC[NH3+]

Specifications

Density 0.851
Boiling Point 114°C to 116°C
Flash Point 41°C (105°F)
Odor Amine-like
Refractive Index 1.4395
Linear Formula H2NCH2CH2NHCH3
Quantity 100 g
UN Number UN2734
Beilstein 878141
Sensitivity Air sensitive; Hygroscopic
Formula Weight 74.13
Percent Purity 95%
Chemical Name or Material N-Methylethylenediamine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 203-707-4
RTECSNumber KV5250500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only