missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1571236

 View more versions of this product

Catalog No. AAL1571236


Only null left
Explore available promotions

Description

Description

N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

N-Methyldiethanolamine
-21°C
243°C
Light
1.469
500 g
n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine
CRVGTESFCCXCTH-UHFFFAOYSA-N
2-[2-hydroxyethyl(methyl)amino]ethanol
7767
≥98%
105-59-9
1.04
127°C (260°F)
C5H13NO2
MFCD00002848
1734441
Soluble in water.
CN(CCO)CCO
119.16
119.17

Chemical Identifiers

105-59-9
119.16
CRVGTESFCCXCTH-UHFFFAOYSA-N
7767
CN(CCO)CCO
C5H13NO2
MFCD00002848
n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine
2-[2-hydroxyethyl(methyl)amino]ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 203-312-7

RTECSNumber : KL7525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only