missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1571222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1571222 100 g
AAL1571236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1571222 Supplier Thermo Scientific Chemicals Supplier No. L1571222

N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyldiethanolamine is a reagent used for protection of boronic acids as N-methyl-O,O-diethanolamine esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 105-59-9
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00002848
InChI Key CRVGTESFCCXCTH-UHFFFAOYSA-N
Synonym n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine
PubChem CID 7767
IUPAC Name 2-[2-hydroxyethyl(methyl)amino]ethanol
SMILES CN(CCO)CCO

Specifications

Melting Point -21°C
Density 1.04
Boiling Point 243°C
Flash Point 127°C (260°F)
Odor Light
Refractive Index 1.469
Quantity 100 g
Beilstein 1734441
Solubility Information Soluble in water.
Formula Weight 119.17
Percent Purity ≥98%
Chemical Name or Material N-Methyldiethanolamine
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 203-312-7
RTECSNumber KL7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only