Learn More
N-Methylcarbazole, 99%, Thermo Scientific Chemicals
$186.73 - $186.73
Chemical Identifiers
| CAS | 1484-12-4 |
|---|---|
| Molecular Formula | C13H11N |
| Molecular Weight (g/mol) | 181.24 |
| InChI Key | SDFLTYHTFPTIGX-UHFFFAOYSA-N |
| Synonym | n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole |
| PubChem CID | 15142 |
| IUPAC Name | 9-methylcarbazole |
| SMILES | CN1C2=CC=CC=C2C3=CC=CC=C31 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC441050050
|
Thermo Scientific Chemicals
441050050 |
5 g |
Each for $186.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1484-12-4 | |
| 181.24 | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| 9-methylcarbazole |
| C13H11N | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 15142 | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 |
Specifications
| 1484-12-4 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| 5 g | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 9-methylcarbazole | |
| 15142 | |
| 99% | |
| N-Methylcarbazole |
| 98.5 | |
| 86.0°C to 89.0°C | |
| 163.0°C to 167.0°C (0.4 Torr) | |
| 98.5% min. (HPLC) | |
| C13H11N | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 | |
| 181.24 | |
| 181.24 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-054-5
RUO – Research Use Only