missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzamide, 99%

Catalog No. p-7023968
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1805706 5 g
AAA1805714 25 g
AAA1805722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1805706 Supplier Thermo Scientific Chemicals Supplier No. A1805706
Only null left

CAS: 613-93-4 | C8H9NO | 135.17 g/mol

It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 613-93-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00011642
InChI Key NCCHARWOCKOHIH-UHFFFAOYSA-N
Synonym benzamide, n-methyl, n-methylbenzenamide, n-methyl-benzamide, n-methylbenzenecarboxamide, unii-k3ed781e08, ccris 4670, n-methyl benzamide, n-methylbenzimidic acid, acmc-1bb7i, dsstox_cid_5570
PubChem CID 11954
IUPAC Name N-methylbenzamide
SMILES CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 76°C to 81°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Methylbenzamide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-362-3
RTECSNumber CV5570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only