missing translation for 'onlineSavingsMsg'
Learn More

N-Methylanilinium trifluoroacetate, 99%

Catalog No. AAA1778122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1778122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1778122 Supplier Thermo Scientific Chemicals Supplier No. A1778122
Only null left

CAS: 29885-95-8 | C7H10N·CF3CO2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29885-95-8
Molecular Formula C7H10N·CF3CO2
MDL Number MFCD00043278
Synonym TAMA; Trifluoroacetic acid N-methylanilinium salt

Specifications

Melting Point 64°C to 68°C
Quantity 100 g
UN Number UN3077
Beilstein 3638133
Sensitivity Hygroscopic
Formula Weight 221.18
Percent Purity 99%
Chemical Name or Material N-Methylanilinium trifluoroacetate
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H311-H373-H332
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 249-927-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only