missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(tetrahydrothiopyran-4-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 950603-22-2
Molecular Formula C7H15NS
Molecular Weight (g/mol) 145.264
MDL Number MFCD09879924
InChI Key FPKJBESBCAZWPI-UHFFFAOYSA-N
Synonym methyl thian-4-ylmethyl amine, n-methyl tetrahydrothiopyran-4-yl methylamine, n-methyl tetrahydro-2h-thiopyran-4-yl methylamine, methyl thian-4-yl methyl amine, n-methyl-1-thian-4-yl methanamine, n-methyl-n-tetrahydro-2h-thiopyran-4-ylmethyl amine
PubChem CID 24229643
IUPAC Name N-methyl-1-(thian-4-yl)methanamine
SMILES CNCC1CCSCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC61946CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61946DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

950603-22-2
145.264
FPKJBESBCAZWPI-UHFFFAOYSA-N
24229643
CNCC1CCSCC1
C7H15NS
MFCD09879924
methyl thian-4-ylmethyl amine, n-methyl tetrahydrothiopyran-4-yl methylamine, n-methyl tetrahydro-2h-thiopyran-4-yl methylamine, methyl thian-4-yl methyl amine, n-methyl-1-thian-4-yl methanamine, n-methyl-n-tetrahydro-2h-thiopyran-4-ylmethyl amine
N-methyl-1-(thian-4-yl)methanamine

Specifications

950603-22-2
100.0
C7H15NS
methyl thian-4-ylmethyl amine, n-methyl tetrahydrothiopyran-4-yl methylamine, n-methyl tetrahydro-2h-thiopyran-4-yl methylamine, methyl thian-4-yl methyl amine, n-methyl-1-thian-4-yl methanamine, n-methyl-n-tetrahydro-2h-thiopyran-4-ylmethyl amine
CNCC1CCSCC1
145.264
145.27
N-Methyl(tetrahydrothiopyran-4-yl)methylamine
96.5
97%
MFCD09879924
FPKJBESBCAZWPI-UHFFFAOYSA-N
N-methyl-1-(thian-4-yl)methanamine
24229643
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents