missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(tetrahydropyran-4-ylmethyl)amine, 97%, Thermo Scientific™

Catalog No. p-3881757
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29946DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29946DA Supplier Thermo Scientific Chemicals Supplier No. CC29946DA

Chemical Identifiers

CAS 439081-52-4
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.20
MDL Number MFCD06739009
InChI Key WMBCUXKYKVTJRF-UHFFFAOYSA-N
Synonym methyl oxan-4-ylmethyl amine, methyl-tetrahydropyran-4-ylmethyl amine, n-methyl-1-tetrahydro-2h-pyran-4-yl methanamine, methyl-tetrahydro-pyran-4-ylmethyl-amine, n-methyl-tetrahydropyran-4-ylmethyl amine, 4-methylamino methyl tetrahydropyran, 2h-pyran-4-methanamine, tetrahydro-n-methyl, n-methyl-1-tetrahydro-2h-pyran-4-yl methylamine, n-methyl tetrahydro-2h-pyran-4-yl methanamine, 4-methylamino methyl tetrahydro-2h-pyran
PubChem CID 22225668
IUPAC Name N-methyl-1-(oxan-4-yl)methanamine
SMILES CNCC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 129.2
Percent Purity 97%
Chemical Name or Material N-methyl-(tetrahydropyran-4-ylmethyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.