missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-tert-butylamine, 97%

Catalog No. p-2691951
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368810250 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC368810250 Supplier Thermo Scientific Chemicals Supplier No. 368810250
Only null left

CAS: 14610-37-8 | C5H13N | 87.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14610-37-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.16
MDL Number MFCD00042853
InChI Key ZQGJEUVBUVKZKS-UHFFFAOYSA-N
Synonym n-methyl-tert-butylamine, n-tert-butylmethylamine, 2-propanamine, n,2-dimethyl, tert-butylmethylamine, tert-butyl methyl amine, n-methyl-tert.-butylamine, 2-propanamine,n,2-dimethyl, t-butylmethylamine, methyl-t-butylamine, n-t-butylmethylamine
PubChem CID 84552
IUPAC Name N,2-dimethylpropan-2-amine
SMILES CC(C)(C)NC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.7270g/mL
Boiling Point 67.0°C to 69.0°C
Flash Point −26°C
Assay Percent Range 96% (GC)
Packaging Glass Bottle
Refractive Index 1.3950 to 1.3970
Linear Formula (CH3)3CNHCH3
Quantity 25 mL
Beilstein 04,IV,659
Specific Gravity 0.727
Formula Weight 87.16
Percent Purity 97%
Chemical Name or Material N-Methyl-tert-butylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 238-646-2

RUO – Research Use Only