missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Paroxetine 98.0+%, TCI America™

Catalog No. M2645200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2645200MG 200 mg
M26451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2645200MG Supplier TCI America Supplier No. M2645200MG

Chemical Identifiers

CAS 110429-36-2
Molecular Formula C20H22FNO3
Molecular Weight (g/mol) 343.40
MDL Number MFCD03788781
InChI Key MOJZPKOBKCXNKG-UHFFFAOYNA-N
PubChem CID 10947895
IUPAC Name 3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine
SMILES CN1CCC(C(COC2=CC3=C(OCO3)C=C2)C1)C1=CC=C(F)C=C1

Specifications

Melting Point 111°C
Color White
Quantity 200 mg
Formula Weight 343.40
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl Paroxetine
TSCA No
Recommended Storage Freezer