missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-trimethylsilyltrifluoroacetamide 95.0+%, TCI America™

Catalog No. M06725ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06725ML 5 mL
M067225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06725ML Supplier TCI America Supplier No. M06725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 1993
Formula Weight 199.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-trimethylsilyltrifluoroacetamide [Trimethylsilylating Agent]
RTECSNumber AC9800000
TSCA Yes