missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-trimethylsilylacetamide 90.0+%, TCI America™

Catalog No. M053610G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053610G 10 g
M053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M053610G Supplier TCI America Supplier No. M053610G

Chemical Identifiers

CAS 7449-74-3
Molecular Formula C6H15NOSi
Molecular Weight (g/mol) 145.277
MDL Number MFCD00040758
InChI Key QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonym n-methyl-n-trimethylsilyl acetamide, acetamide, n-methyl-n-trimethylsilyl, n-trimethylsilyl-n-methylacetamide, n-methyl-n-tms-acetamide, acmc-1bei6, qhuobldkfgcvcg-uhfffaoysa, n-methylacetamide, tms derivative, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
PubChem CID 81953
IUPAC Name N-methyl-N-trimethylsilylacetamide
SMILES CC(=O)N(C)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 154°C
Quantity 10 g
UN Number 2924
Formula Weight 145.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-trimethylsilylacetamide [Trimethylsilylating Agent]
TSCA Yes
Recommended Storage Refrigerator