missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide synthesis grade, ≥98%, MilliporeSigma™ Supelco™
N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a derivatizing reagent. It can be used as alternatives to diazomethane.
$243.46 - $686.99
Chemical Identifiers
| MDL Number | MFCD00000411 |
|---|---|
| Synonym | N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA |
Chemical Identifiers
| MFCD00000411 | |
| N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA |
Specifications
| 24589-78-4 | |
| 130°C to 132°C | |
| Ampule | |
| MFCD00000411 | |
| N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA | |
| 199.25 | |
| ≥98% |
| 1.075 g/mL (at 25°C) | |
| 26°C - Closed Cup | |
| CF3 CON(CH3)Si(CH3)3 | |
| 5 mL | |
| >1 (vs. air) | |
| 8.8 mmHg (27°C) | |
| Synthesis Grade |
Safety and Handling
RTECSNumber : AC9800000