missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%

Catalog No. AC221580050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221580050 5 g Glass bottle
AC221580250 25 g Glass bottle
AC221581000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221580050 Supplier Thermo Scientific Chemicals Supplier No. 221580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24589-78-4 | C6H12F3NOSi | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, Silylation reagent

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 130°C to 132°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3782 to 1.3802
Linear Formula CF3CON(CH3)Si(CH3)3
Quantity 5 g
Specific Gravity 1.07
Solubility Information Solubility in water: reacts
Formula Weight 199.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 246-331-6

RUO – Research Use Only