missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-n-octylamine, 97+%

Catalog No. p-7047306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1863906 5 g
AAL1863914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1863906 Supplier Thermo Scientific Chemicals Supplier No. L1863906
Only null left

CAS: 2439-54-5 | C9H21N | 143.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2439-54-5
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD00048927
InChI Key SEGJNMCIMOLEDM-UHFFFAOYSA-N
Synonym n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
PubChem CID 75538
IUPAC Name N-methyloctan-1-amine
SMILES CCCCCCCCNC

Specifications

Density 0.79
Boiling Point 68°C (8 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.427
Linear Formula CH3(CH2)7NHCH3
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 143.27
Percent Purity 98%
Chemical Name or Material N-Methyl-n-octylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 219-462-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only