missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-methyl-3-phenylisoxazol-4-yl)methyl]amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 857283-57-9
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.26
MDL Number MFCD07772789
InChI Key IEMWBRRLWLUKQI-UHFFFAOYSA-N
Synonym n-methyl-n-5-methyl-3-phenylisoxazol-4-yl methyl amine, 4-isoxazolemethanamine,n,5-dimethyl-3-phenyl, methyl 5-methyl-3-phenyl-1,2-oxazol-4-yl methyl amine, n-methyl-1-5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, methyl 5-methyl-3-phenylisoxazol-4-yl methyl amine
PubChem CID 7162097
IUPAC Name N-methyl-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
SMILES CNCC1=C(C)ON=C1C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC00546DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

857283-57-9
202.26
IEMWBRRLWLUKQI-UHFFFAOYSA-N
7162097
CNCC1=C(C)ON=C1C1=CC=CC=C1
C12H14N2O
MFCD07772789
n-methyl-n-5-methyl-3-phenylisoxazol-4-yl methyl amine, 4-isoxazolemethanamine,n,5-dimethyl-3-phenyl, methyl 5-methyl-3-phenyl-1,2-oxazol-4-yl methyl amine, n-methyl-1-5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, methyl 5-methyl-3-phenylisoxazol-4-yl methyl amine
N-methyl-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine

Specifications

857283-57-9
100.0
C12H14N2O
n-methyl-n-5-methyl-3-phenylisoxazol-4-yl methyl amine, 4-isoxazolemethanamine,n,5-dimethyl-3-phenyl, methyl 5-methyl-3-phenyl-1,2-oxazol-4-yl methyl amine, n-methyl-1-5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, methyl 5-methyl-3-phenylisoxazol-4-yl methyl amine
CNCC1=C(C)ON=C1C1=CC=CC=C1
202.26
202.26
N-methyl-N-[(5-methyl-3-phenylisoxazol-4-yl)methyl]amine
96.5
97%
MFCD07772789
IEMWBRRLWLUKQI-UHFFFAOYSA-N
N-methyl-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
7162097
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents