missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-((4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl)amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC29246DA

 View more versions of this product

Catalog No. CC29246DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-methyl-N-[(4-methyl-2-phenyl-1, 3-thiazol-5-yl)methyl]amine
96.5
97%
C12H14N2S
1g
GFAGRBRYZWAUSV-UHFFFAOYSA-N
N-methyl-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanamine
4961253
97%
850375-02-9
100.0
Amber glass bottle
MFCD07368511
n-methyl-n-4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-thiazol-5-yl methanamine, methyl 4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, 5-thiazolemethanamine,n,4-dimethyl-2-phenyl, n-methyl-1-4-methyl-2-phenylthiazol-5-yl methanamine, methyl 4-methyl-2-phenyl 1,3-thiazol-5-yl methyl amine, mmj
CC1=C(SC(=N1)C2=CC=CC=C2)CNC
218.318
218.32

Chemical Identifiers

850375-02-9
218.318
GFAGRBRYZWAUSV-UHFFFAOYSA-N
4961253
CC1=C(SC(=N1)C2=CC=CC=C2)CNC
C12H14N2S
MFCD07368511
n-methyl-n-4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-thiazol-5-yl methanamine, methyl 4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, 5-thiazolemethanamine,n,4-dimethyl-2-phenyl, n-methyl-1-4-methyl-2-phenylthiazol-5-yl methanamine, methyl 4-methyl-2-phenyl 1,3-thiazol-5-yl methyl amine, mmj
N-methyl-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions