missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 879896-58-9
Molecular Formula C12H19N3
Molecular Weight (g/mol) 205.305
MDL Number MFCD09025859
InChI Key CPEUGLUNQVAGDV-UHFFFAOYSA-N
Synonym n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
PubChem CID 17851834
IUPAC Name N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine
SMILES CNCC1=CC(=NC=C1)N2CCCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC50346CB
SDS View Documents
Thermo Scientific Chemicals
CC50346CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50346DA
SDS View Documents
Thermo Scientific Chemicals
CC50346DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

879896-58-9
205.305
CPEUGLUNQVAGDV-UHFFFAOYSA-N
17851834
CNCC1=CC(=NC=C1)N2CCCCC2
C12H19N3
MFCD09025859
n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine

Specifications

879896-58-9
100.0
Amber Glass Bottle
MFCD09025859
n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
CNCC1=CC(=NC=C1)N2CCCCC2
205.305
205.3
N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine
96.5
97%
C12H19N3
250mg
CPEUGLUNQVAGDV-UHFFFAOYSA-N
N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine
17851834
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents