missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-({2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methyl)amine, 97%, Thermo Scientific™

Catalog No. CC46046CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46046CB 250mg Amber Glass Bottle
CC46046DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC46046CB Supplier Thermo Scientific Chemicals Supplier No. CC46046CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 857284-26-5
Molecular Formula C12H11F3N2S
Molecular Weight (g/mol) 272.289
MDL Number MFCD07772873
InChI Key IBCDDBBGPGTKNC-UHFFFAOYSA-N
Synonym n-methyl-1-2-4-trifluoromethyl phenyl-1,3-thiazol-4-yl methanamine, n-methyl-n-2-4-trifluoromethyl phenyl-1,3-thiazol-4-yl methylamine, n-methyl-n-2-4-trifluoromethyl phenyl-1,3-thiazol-4-yl methyl amine, methyl 2-4-trifluoromethyl phenyl-1,3-thiazol-4-yl methyl amine, n-methyl-1-2-4-trifluoromethyl phenyl-4-thiazolyl methanamine
PubChem CID 7164660
IUPAC Name N-methyl-1-[2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl]methanamine
SMILES CNCC1=CSC(=N1)C2=CC=C(C=C2)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 272.29
Percent Purity 97%
Chemical Name or Material N-methyl-N-({2-[4-(trifluoromethyl)phenyl]-1, 3-thiazol-4-yl}methyl)amine
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.