missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine, 97%, Thermo Scientific™

Catalog No. p-3486388
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC44246DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC44246DA Supplier Thermo Scientific Chemicals Supplier No. CC44246DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 884507-20-4
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD08690256
InChI Key ODIDITAYVOGYIV-UHFFFAOYSA-N
Synonym n-methyl-n-1-methyl-1h-indol-6-yl methyl amine, methyl 1-methylindol-6-yl methyl amine, 1h-indole-6-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-6-yl methanamine, 1h-indole-6-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-6-ylmethyl-amine
PubChem CID 18525770
IUPAC Name N-methyl-1-(1-methylindol-6-yl)methanamine
SMILES CNCC1=CC2=C(C=C1)C=CN2C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 174.25
Percent Purity 95%
Chemical Name or Material N-methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.