missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-prolinol, 96%

Catalog No. p-7041557
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6257306 5 g
AAH6257314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6257306 Supplier Thermo Scientific Chemicals Supplier No. H6257306

CAS: 34381-71-0 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34381-71-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00011727
InChI Key VCOJPHPOVDIRJK-UHFFFAOYNA-N
Synonym n-methyl-l-prolinol, s---1-methyl-2-pyrrolidinemethanol, n-methylprolinol, 2s-1-methylpyrrolidin-2-yl methanol, s-1-methylpyrrolidin-2-yl methanol, 2-pyrrolidinemethanol, 1-methyl-, 2s, 2s-1-methyl-2-pyrrolidinyl methanol, s---2-hydroxymethyl-1-methylpyrrolidine, 1-methyl-2-pyrrolidinyl methanol #, n-methyl prolinol
PubChem CID 643492
SMILES CN1CCCC1CO

Specifications

Refractive Index 1.469
Density 0.968
Boiling Point 67°C to 69°C (12mmHg)
Quantity 5 g
Flash Point 63°C (145°F)
Optical Rotation −49° (c=5 in Methanol)
Beilstein 79851
IUPAC Name (1-methylpyrrolidin-2-yl)methanol
Formula Weight 115.17
Percent Purity 96%
Chemical Name or Material N-Methyl-L-prolinol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 251-981-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only