missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-alanine 98.0+%, TCI America™

Catalog No. p-7133220
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24361G 1 g
M24365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24361G Supplier TCI America Supplier No. M24361G

Chemical Identifiers

CAS 3913-67-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00037241
InChI Key GDFAOVXKHJXLEI-VKHMYHEASA-N
Synonym n-methyl-l-alanine, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine, alanine, n-methyl-, l
PubChem CID 5288725
ChEBI CHEBI:17519
IUPAC Name (2S)-2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Melting Point 300°C
Color White
Quantity 1 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-L-alanine