missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-DL-alanine 98.0+%, TCI America™

Catalog No. M0777100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0777100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0777100MG Supplier TCI America Supplier No. M0777100MG

Chemical Identifiers

CAS 600-21-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00063136
InChI Key GDFAOVXKHJXLEI-UHFFFAOYSA-N
Synonym n-methyl-dl-alanine, 2-methylamino propanoic acid, h-n-me-dl-ala-oh, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, alanine, n-methyl-, l, dl-n-methylalanine, me-dl-ala-oh, n-methyl cdl-alanine, n-, a-methyl-dl-alanine
PubChem CID 4377
IUPAC Name 2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Melting Point 317°C
Color White
Quantity 100 mg
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-DL-alanine