missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-DL-alanine, 98%

Catalog No. p-7056399
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6528714 25 g
AAH6528722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6528714 Supplier Thermo Scientific Chemicals Supplier No. H6528714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 600-21-5 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 600-21-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00063136
InChI Key GDFAOVXKHJXLEI-UHFFFAOYSA-N
Synonym n-methyl-dl-alanine, 2-methylamino propanoic acid, h-n-me-dl-ala-oh, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, alanine, n-methyl-, l, dl-n-methylalanine, me-dl-ala-oh, n-methyl cdl-alanine, n-, a-methyl-dl-alanine
PubChem CID 4377
IUPAC Name 2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-DL-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only