missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(chroman-8-ylmethyl)amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1048970-16-6
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD12198126
InChI Key VVCGFZWBAXPSIO-UHFFFAOYSA-N
Synonym n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
PubChem CID 43811059
IUPAC Name 1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine
SMILES CNCC1=CC=CC2=C1OCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC76146CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC76146DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1048970-16-6
177.247
VVCGFZWBAXPSIO-UHFFFAOYSA-N
43811059
CNCC1=CC=CC2=C1OCCC2
C11H15NO
MFCD12198126
n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine

Specifications

1048970-16-6
100.0
C11H15NO
n-methyl-chroman-8-ylmethyl amine, 3,4-dihydro-2h-1-benzopyran-8-ylmethyl methyl amine, chroman-8-ylmethyl methylamine, 1-3,4-dihydro-2h-1-benzopyran-8-yl-n-methylmethanamine
CNCC1=CC=CC2=C1OCCC2
177.247
177.25
N-methyl-(chroman-8-ylmethyl)amine
96.5
97%
MFCD12198126
VVCGFZWBAXPSIO-UHFFFAOYSA-N
1-(3,4-dihydro-2H-chromen-8-yl)-N-methylmethanamine
43811059
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents