missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-bis(trifluoroacetamide), ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187720
Change view
Click to view available options
Quantity:
5 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018471 5 mL
111018296 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018471 Supplier Merck Emd Millipore Supplier No. M07895ML
Only null left

Chemical Identifiers

MDL Number MFCD00000412
Synonym MBTFA (N-methyl-bis(trifluoroacetamide)or BTFA (bis(TFA)); MBTFA

Specifications

Density 1.547 g/mL (at 25°C)
CAS 685-27-8
Boiling Point 121°C to 122°C
Flash Point 45°C
Packaging Glass Bottle
Linear Formula CF3 CON(CH3)COCF3
Quantity 5 mL
Formula Weight 223.07
Percent Purity ≥97.0% (GC)
Grade For GC derivatization