missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-beta-alaninenitrile, 98%

Catalog No. AC126220050
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126220050 5 g Glass bottle
AC126220500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126220050 Supplier Thermo Scientific Chemicals Supplier No. 126220050

CAS: 693-05-0 | C4H8N2 | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-05-0
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001954
InChI Key UNIJBMUBHBAUET-UHFFFAOYSA-N
Synonym 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile
PubChem CID 69656
IUPAC Name 3-(methylamino)propanenitrile
SMILES CNCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.8900g/mL
Boiling Point 182°C to 186°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4300 to 1.432
Linear Formula CH3NHCH2CH2CN
Quantity 5 g
Specific Gravity 0.89
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methyl-β-alaninenitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-740-0
RTECSNumber TZ4910000
TSCA TSCA

RUO – Research Use Only