missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 910036-99-6
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.288
MDL Number MFCD09879908
InChI Key CWRQCHDTRDIGCU-UHFFFAOYSA-N
Synonym n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
PubChem CID 24229584
IUPAC Name N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine
SMILES CNCC1=CN=C(C=C1)OC2CCOCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC58346CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58346DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

910036-99-6
222.288
CWRQCHDTRDIGCU-UHFFFAOYSA-N
24229584
CNCC1=CN=C(C=C1)OC2CCOCC2
C12H18N2O2
MFCD09879908
n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine

Specifications

910036-99-6
100.0
C12H18N2O2
n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
CNCC1=CN=C(C=C1)OC2CCOCC2
222.288
222.29
N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine
96.5
95%
MFCD09879908
CWRQCHDTRDIGCU-UHFFFAOYSA-N
N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine
24229584
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents