Learn More
n-methyl-(5-pyrid-4-ylthien-2-yl)methylamine, 97%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals CC67046CB
Quantity 250mg
Packaging Amber Glass Bottle
 View more versions of this product
Catalog No. CC67046CB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Chemical Identifiers
934570-47-5
204.29
JSAZQLBHWDPNPA-UHFFFAOYSA-N
24229745
CNCC1=CC=C(S1)C1=CC=NC=C1
C11H12N2S
MFCD09879964
n-methyl-5-pyrid-4-ylthien-2-yl methylamine, methyl 5-pyridin-4-yl thiophen-2-yl methyl amine, methyl 5-4-pyridyl 2-thienyl methyl amine, n-methyl-1-5-pyridin-4-yl thiophen-2-yl methanamine, n-methyl-5-pyridin-4-ylthiophen-2-yl methylamine
N-methyl-1-(5-pyridin-4-ylthiophen-2-yl)methanamine
Specifications
N-methyl-(5-pyrid-4-ylthien-2-yl)methylamine
96.5
97%
C11H12N2S
MFCD09879964
JSAZQLBHWDPNPA-UHFFFAOYSA-N
N-methyl-1-(5-pyridin-4-ylthiophen-2-yl)methanamine
24229745
97%
934570-47-5
100.0
Amber Glass Bottle
250mg
n-methyl-5-pyrid-4-ylthien-2-yl methylamine, methyl 5-pyridin-4-yl thiophen-2-yl methyl amine, methyl 5-4-pyridyl 2-thienyl methyl amine, n-methyl-1-5-pyridin-4-yl thiophen-2-yl methanamine, n-methyl-5-pyridin-4-ylthiophen-2-yl methylamine
CNCC1=CC=C(S1)C1=CC=NC=C1
204.29
204.3
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger