missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(4-phenyltetrahydropyran-4-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC69146DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC69146DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

958443-30-6
205.301
OHCOJCIAAGLEHJ-UHFFFAOYSA-N
33589539
CNCC1(CCOCC1)C2=CC=CC=C2
C13H19NO
MFCD11841073
n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
N-methyl-1-(4-phenyloxan-4-yl)methanamine

Specifications

N-methyl-(4-phenyltetrahydropyran-4-yl)methylamine
96.5
97%
C13H19NO
MFCD11841073
OHCOJCIAAGLEHJ-UHFFFAOYSA-N
N-methyl-1-(4-phenyloxan-4-yl)methanamine
33589539
97%
958443-30-6
100.0
Amber Glass Bottle
1g
n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
CNCC1(CCOCC1)C2=CC=CC=C2
205.301
205.3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents