missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(4-phenyltetrahydropyran-4-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC69146CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69146CB 250mg Amber Glass Bottle
CC69146DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC69146CB Supplier Thermo Scientific Chemicals Supplier No. CC69146CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 958443-30-6
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD11841073
InChI Key OHCOJCIAAGLEHJ-UHFFFAOYSA-N
Synonym n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
PubChem CID 33589539
IUPAC Name N-methyl-1-(4-phenyloxan-4-yl)methanamine
SMILES CNCC1(CCOCC1)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 205.3
Percent Purity 97%
Chemical Name or Material N-methyl-(4-phenyltetrahydropyran-4-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.