missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 921938-85-4
Molecular Formula C10H15N3O
Molecular Weight (g/mol) 193.25
MDL Number MFCD09817509
InChI Key SMECUBWISXUYSV-UHFFFAOYSA-N
Synonym n-methyl 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, methyl 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methyl amine, n-methyl-1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, methyl 4-methyl 2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methyl amine, n-methyl-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, n-methyl-1-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine
PubChem CID 24229654
IUPAC Name N-methyl-1-(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine
SMILES CNCC1=CC2=C(N=C1)N(C)CCO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC62046CB
SDS View Documents
Thermo Scientific Chemicals
CC62046CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62046DA
SDS View Documents
Thermo Scientific Chemicals
CC62046DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

921938-85-4
193.25
SMECUBWISXUYSV-UHFFFAOYSA-N
24229654
CNCC1=CC2=C(N=C1)N(C)CCO2
C10H15N3O
MFCD09817509
n-methyl 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, methyl 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methyl amine, n-methyl-1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, methyl 4-methyl 2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methyl amine, n-methyl-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, n-methyl-1-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine
N-methyl-1-(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine

Specifications

921938-85-4
100.0
Amber Glass Bottle
MFCD09817509
n-methyl 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, methyl 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methyl amine, n-methyl-1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, methyl 4-methyl 2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methyl amine, n-methyl-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, n-methyl-1-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine
CNCC1=CC2=C(N=C1)N(C)CCO2
193.25
193.25
N-Methyl(4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine
96.5
95%
C10H15N3O
250mg
SMECUBWISXUYSV-UHFFFAOYSA-N
N-methyl-1-(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine
24229654
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'