missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Thermo Scientific™

Catalog No. CC18146CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC18146CB 250mg Amber Glass Bottle
CC18146DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC18146CB Supplier Thermo Scientific Chemicals Supplier No. CC18146CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1031843-28-3
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD11109322
InChI Key XCBYKJIAOCVNGY-UHFFFAOYSA-N
Synonym n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine
PubChem CID 33589464
IUPAC Name N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine
SMILES CC1=C(OC(=N1)C2=CC=CC=C2)CNC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material N-Methyl-(4-methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.