missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride, Tech., Thermo Scientific™

Catalog No. p-3586866
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC54296CB 250mg Amber Glass Bottle
CC54296DA 1g Amber Glass Bottle
CC54296DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC54296CB Supplier Thermo Scientific Chemicals Supplier No. CC54296CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 944450-82-2
Molecular Formula C6H9BrClNS
Molecular Weight (g/mol) 242.559
MDL Number MFCD09817483
InChI Key GORGMQYTNXIOBQ-UHFFFAOYSA-N
Synonym n-methyl-4-bromothien-3-yl methyl amine hydrochloride, 1-4-bromothiophen-3-yl-n-methylmethanamine hydrochloride, 4-bromothiophen-3-yl methyl methyl amine hydrochloride, 4-bromo 3-thienyl methyl methylamine, chloride, 4-bromo-thiophen-3-ylmethyl-methyl-amine hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride tech, 1-4-bromothiophen-3-yl-n-methylmethanamine-hydrogen chloride 1/1
PubChem CID 24229540
IUPAC Name 1-(4-bromothiophen-3-yl)-N-methylmethanamine;hydrochloride
SMILES CNCC1=CSC=C1Br.Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 242.57
Percent Purity Tech.
Grade Technical
Chemical Name or Material N-methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.