missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(1-methyl-1H-pyrazol-3-yl)benzylamine, 90%, Thermo Scientific™

Catalog No. cc23846cb
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23846CB 250mg Amber Glass Bottle
CC23846DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC23846CB Supplier Thermo Scientific Chemicals Supplier No. CC23846CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 179873-47-3
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.273
MDL Number MFCD09702353
InChI Key BBOCGPKUAXCMLG-UHFFFAOYSA-N
Synonym n-methyl-4-1-methyl-1h-pyrazol-3-yl benzylamine, benzenemethanamine,n-methyl-4-1-methyl-1h-pyrazol-3-yl, methyl 4-1-methylpyrazol-3-yl phenyl methyl amine, n-methyl-1-4-1-methyl-1h-pyrazol-3-yl phenyl methanamine, methyl 4-1-methyl-1h-pyrazol-3-yl phenyl methyl amine
PubChem CID 24229474
IUPAC Name N-methyl-1-[4-(1-methylpyrazol-3-yl)phenyl]methanamine
SMILES CNCC1=CC=C(C=C1)C2=NN(C=C2)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 201.27
Percent Purity 90%
Chemical Name or Material N-methyl-4-(1-methyl-1H-pyrazol-3-yl)benzylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.