missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(3-methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™

Catalog No. CC27046DA
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27046DA 1g Amber Glass Bottle
CC27046CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC27046DA Supplier Thermo Scientific Chemicals Supplier No. CC27046DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 401647-22-1
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.16
MDL Number MFCD08060065
InChI Key UQKWKRSUELRKSK-UHFFFAOYSA-N
Synonym n-methyl-1-3-methylisoxazol-5-yl methanamine, n-methyl-3-methylisoxazol-5-ylmethyl amine, methyl 3-methyl-1,2-oxazol-5-yl methyl amine, n-methyl 3-methylisoxazol-5-yl methanamine, methyl 3-methylisoxazol-5-yl methyl amine, n-methyl-1-3-methyl-1,2-oxazol-5-yl methanamine, 5-isoxazolemethanamine,n,3-dimethyl, 5-isoxazolemethanamine, n,3-dimethyl, methyl 3-methylisoxazol-5-ylmethyl amine, n-methyl-n-3-methylisoxazol-5-yl methyl amine
PubChem CID 25220779
IUPAC Name N-methyl-1-(3-methyl-1,2-oxazol-5-yl)methanamine
SMILES CNCC1=CC(C)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material N-methyl-(3-methylisoxazol-5-ylmethyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.