Learn More
n-methyl(3-methyl-2-furyl)methylamine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC55546DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 916766-86-4 | |
| 125.171 | |
| KBNPUPCSMGGUHM-UHFFFAOYSA-N | |
| 24229550 | |
| CC1=C(OC=C1)CNC |
| C7H11NO | |
| MFCD09879907 | |
| methyl 3-methylfuran-2-yl methyl amine, n-methyl 3-methyl-2-furyl methylamine, n-methyl 3-methylfur-2-yl methylamine, 2-furanmethanamine, n,3-dimethyl, n-methyl-1-3-methylfuran-2-yl methanamine, 3-methyl-2-methylamino methyl furan, methyl 3-methyl 2-furyl methyl amine | |
| N-methyl-1-(3-methylfuran-2-yl)methanamine |
Specifications
| N-methyl(3-methyl-2-furyl)methylamine | |
| 89.5 | |
| 90% | |
| C7H11NO | |
| MFCD09879907 | |
| KBNPUPCSMGGUHM-UHFFFAOYSA-N | |
| N-methyl-1-(3-methylfuran-2-yl)methanamine | |
| 24229550 | |
| 90% |
| 916766-86-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| 1g | |
| methyl 3-methylfuran-2-yl methyl amine, n-methyl 3-methyl-2-furyl methylamine, n-methyl 3-methylfur-2-yl methylamine, 2-furanmethanamine, n,3-dimethyl, n-methyl-1-3-methylfuran-2-yl methanamine, 3-methyl-2-methylamino methyl furan, methyl 3-methyl 2-furyl methyl amine | |
| CC1=C(OC=C1)CNC | |
| 125.171 | |
| 125.17 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger