missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-chloropropylamine Hydrochloride 99.0+%, TCI America™

Catalog No. M10481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10481G 1 g
M10485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10481G Supplier TCI America Supplier No. M10481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97145-88-5
Molecular Formula C4H11Cl2N
Molecular Weight (g/mol) 144.039
MDL Number MFCD00144935
InChI Key SHBWHBGVZIDIQY-UHFFFAOYSA-N
Synonym 3-(Methylamino)propyl Chloride Hydrochloride
PubChem CID 22107021
IUPAC Name 3-chloro-N-methylpropan-1-amine;hydrochloride
SMILES CNCCCCl.Cl

Specifications

Melting Point 105°C
Color White
Quantity 1 g
Formula Weight 144.04
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-3-chloropropylamine Hydrochloride