missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridine-2-yl)ethyl]ethanamine trihydrochloride, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 2095467-43-7 |
| Flash Point | Not Applicable |
| Molecular Formula | C15H19N3 · 3 HCl |
| Linear Formula | C15H19N3 · 3 HCl |
| Quantity | 25 mg |
| Synonym | N-Methyl-N-[2-(2-pyridinyl)ethyl]-2-pyridineethanamine trihydrochloride |
| Formula Weight | 350.71 |
| Grade | BP |