missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-phenylethylamine 97.0+%, TCI America™

Catalog No. M19505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19505G 5 g
M195025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19505G Supplier TCI America Supplier No. M19505G

Chemical Identifiers

CAS 589-08-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008291
InChI Key SASNBVQSOZSTPD-UHFFFAOYSA-N
Synonym n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine
PubChem CID 11503
IUPAC Name N-methyl-2-phenylethanamine
SMILES CNCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 205°C
Quantity 5 g
Formula Weight 135.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-2-phenylethylamine
RTECSNumber SH9625000
TSCA No