missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-(2-methyl-1,3-oxazol-4-yl)methylamine, Thermo Scientific™

Catalog No. CC57946DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC57946DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC57946DA Supplier Thermo Scientific Chemicals Supplier No. CC57946DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1065073-46-2
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.159
InChI Key SFLPQWGHHRKLNJ-UHFFFAOYSA-N
Synonym methyl 2-methyl-1,3-oxazol-4-yl methyl amine, 4-methylaminomethyl-2-methyl-1,3-oxazole, 4-oxazolemethanamine, n,2-dimethyl, 2-methyl-4-methylamino methyl-1,3-oxazole, methyl 2-methyl 1,3-oxazol-4-yl methyl amine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methanamine, n-methyl-1-2-methyl-1,3-oxazol-4-yl methylamine, n-methyl-2-methyl-1,3-oxazol-4-yl methylamine
PubChem CID 44119607
IUPAC Name N-methyl-1-(2-methyl-1,3-oxazol-4-yl)methanamine
SMILES CC1=NC(=CO1)CNC

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material N-methyl-(2-methyl-1, 3-oxazol-4-yl)methylamine
Hazard Category GHS Signal Word: Danger