missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Catalog No. CC49646CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC49646CB 250mg Amber Glass Bottle
CC49646DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC49646CB Supplier Thermo Scientific Chemicals Supplier No. CC49646CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 879896-56-7
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD09702360
InChI Key LJDBGVKOXDSIQQ-UHFFFAOYSA-N
Synonym n-methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, benzenemethanamine,n-methyl-2-3-methyl-1,2,4-oxadiazol-5-yl, methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, methyl 2-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
PubChem CID 24229504
IUPAC Name N-methyl-1-[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
SMILES CNCC1=CC=CC=C1C1=NC(C)=NO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl[2-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.