missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[(2,3-dihydrobenzo[b]furan-7-yl)methyl]amine, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00946DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC00946DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

389845-43-6
163.22
WVSLYVGYBIMHJY-UHFFFAOYSA-N
26343571
CNCC1=CC=CC2=C1OCC2
C10H13NO
MFCD10700046
n-methyl-2,3-dihydrobenzo b furan-7-yl methyl amine, 2,3-dihydro-1-benzofuran-7-ylmethyl methyl amine, 1-2,3-dihydrobenzofuran-7-yl-n-methylmethanamine, 2,3-dihydro-benzofuran-7-ylmethyl methylamine, 7-benzofuranmethanamine,2,3-dihydro-n-methyl, 2,3-dihydrobenzo 2,3-b furan-7-ylmethyl methylamine, 1-2,3-dihydro-1-benzofuran-7-yl-n-methylmethanamine
1-(2,3-dihydro-1-benzofuran-7-yl)-N-methylmethanamine

Specifications

N-methyl-[(2, 3-dihydrobenzo[b]furan-7-yl)methyl]amine
89.5
90%
C10H13NO
MFCD10700046
WVSLYVGYBIMHJY-UHFFFAOYSA-N
1-(2,3-dihydro-1-benzofuran-7-yl)-N-methylmethanamine
26343571
90%
389845-43-6
100.0
Amber Glass Bottle
1g
n-methyl-2,3-dihydrobenzo b furan-7-yl methyl amine, 2,3-dihydro-1-benzofuran-7-ylmethyl methyl amine, 1-2,3-dihydrobenzofuran-7-yl-n-methylmethanamine, 2,3-dihydro-benzofuran-7-ylmethyl methylamine, 7-benzofuranmethanamine,2,3-dihydro-n-methyl, 2,3-dihydrobenzo 2,3-b furan-7-ylmethyl methylamine, 1-2,3-dihydro-1-benzofuran-7-yl-n-methylmethanamine
CNCC1=CC=CC2=C1OCC2
163.22
163.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions