missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-propylamine, 97%

CAS: 627-35-0 | C4H11N | 73.139 g/mol

Supplier:  Thermo Scientific Chemicals A1690306

 View more versions of this product

Catalog No. AAA1690306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methyl-1-propylamine
0.713
−1.1°C (30°F)
C4H11N
5 g
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
CCCNC
73.139
73.14
627-35-0
62°C to 64°C
Amine-like
MFCD00009361
UN2733
GVWISOJSERXQBM-UHFFFAOYSA-N
N-methylpropan-1-amine
12315
97%

Chemical Identifiers

627-35-0
73.139
GVWISOJSERXQBM-UHFFFAOYSA-N
12315
CCCNC
C4H11N
MFCD00009361
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
N-methylpropan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only