missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-propylamine, 97%

Catalog No. AAA1690303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1690303 1 g
AAA1690306 5 g
AAA1690314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1690303 Supplier Thermo Scientific Chemicals Supplier No. A1690303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 627-35-0 | C4H11N | 73.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009361
InChI Key GVWISOJSERXQBM-UHFFFAOYSA-N
Synonym n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
PubChem CID 12315
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC

Specifications

Density 0.713
Boiling Point 62°C to 64°C
Flash Point −1.1°C (30°F)
Odor Amine-like
Quantity 1 g
UN Number UN2733
Formula Weight 73.14
Percent Purity 97%
Chemical Name or Material N-Methyl-1-propylamine
Hazard Category H225-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only